Butyl methyl ether ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 1-Methoxybutane

M0504-25ML M0504-500ML M0504-100ML
TCIAM0504-25MLEA 71.5 EUR
TCIAM0504-25ML TCIAM0504-500ML TCIAM0504-100ML
Butyl methyl ether ≥99.0% (by GC)
Butyl methyl ether
Formule: C₅H₁₂O
Kookpunt: 70°C
Vlampunt: –10°C
MDL Nummer: MFCD00009453
CAS nummer: 628-28-4
EINECS: 211-033-7
UN: 2350
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c764e1377bd777ff5b80c4ee26a171df5fe51e8e&vid=c764e1377bd777ff5b80c4ee26a171df5fe51e8e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR