N-Methyl-N-trimethylsilylacetamide ≥90.0% (by GC) trimethylsilylating agent
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
M0536-10G
M0536-25G
TCIAM0536-10GEA
84.7
EUR
TCIAM0536-10G
TCIAM0536-25G
N-Methyl-N-trimethylsilylacetamide ≥90.0% (by GC) trimethylsilylating agent
N-Methyl-N-trimethylsilylacetamide
Formule:
C₆H₁₅NOSi Kookpunt: 154°C Vlampunt: 48°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00040758 CAS nummer: 7449-74-3 EINECS: 231-217-0 UN: 2924 ADR: 3,III |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 90.0 % |
Effect test of TMS derivatization | Effective as derivatization agent Myristic Acid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...