Maltitol ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M0601-25G M0601-500G
TCIAM0601-25GEA 18.7 EUR
TCIAM0601-25G TCIAM0601-500G
Maltitol ≥95.0% (by GC)
Maltitol
Formule: C₁₂H₂₄O₁₁
MDL Nummer: MFCD00006600
CAS nummer: 585-88-6
EINECS: 209-567-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 145.0 to 152.0 deg-C
Specific rotation [a]20/D +100 to +110 deg (C=10, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR