1-Methyl-3-p-tolyltriazene ≥98.0% (by HPLC, titration analysis) for esterification

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

M0641-1G M0641-25G
TCIAM0641-1GEA 40.7 EUR
TCIAM0641-1G TCIAM0641-25G
1-Methyl-3-p-tolyltriazene ≥98.0% (by HPLC, titration analysis) for esterification
1-Methyl-3-p-tolyltriazene
Formule: C₈H₁₁N₃
MDL Nummer: MFCD00008282
CAS nummer: 21124-13-0
EINECS: 244-223-3
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) 98.0 to 102.0 %
Effect test of ester derivatization Effective as derivatization agent n-Octanoic Acid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR