Diisopropyl malonate ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Malonic Acid Diisopropyl Ester
M0648-25ML M0648-500ML
TCIAM0648-25MLEA 17.6 EUR
TCIAM0648-25ML TCIAM0648-500ML
Diisopropyl malonate ≥99.0% (by GC)
Diisopropyl malonate
Formule: C₉H₁₆O₄
MW: 188.22 g/mol
Kookpunt: 106°C
Smeltpunt: –51°C
Vlampunt: 95°C
MDL Nummer: MFCD00059359
CAS nummer: 13195-64-7
EINECS: 236-156-3
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %