N-Methylacetoacetamide ∼70% in water

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: N-Acetoacetylmethylamine

M0657-25G M0657-500G
TCIAM0657-25GEA 17.6 EUR
TCIAM0657-25G TCIAM0657-500G
N-Methylacetoacetamide ∼70% in water
N-Methylacetoacetamide
Formule: C₅H₉NO₂
Vlampunt: 105°C
MDL Nummer: MFCD00044026
CAS nummer: 20306-75-6
EINECS: 243-723-9

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Concentration (with Total Nitrogen) 70.0 to 77.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR