N-Methyl-D-glucamine hydrochloride ≥98.0% (by HPLC, titration analysis) for buffer

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: N-Methyl-D-glucamine Hydrochloride

M0713-25G M0713-5G
TCIAM0713-25GEA 182 EUR
TCIAM0713-25G TCIAM0713-5G
N-Methyl-D-glucamine hydrochloride ≥98.0% (by HPLC, titration analysis) for buffer
N-Methyl-D-glucamine hydrochloride
Formule: C₇H₁₇NO₅
MDL Nummer: MFCD00060164
CAS nummer: 35564-86-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 148.0 to 153.0 deg-C
Specific rotation [a]20/D -19.0 to -22.0 deg (C=10, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR