3-Methyl-2-oxazolidone ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M0781-25G
M0781-500G
M0781-100G
TCIAM0781-25GEA
44
EUR
TCIAM0781-25G
TCIAM0781-500G
TCIAM0781-100G
3-Methyl-2-oxazolidone ≥99.0% (by GC)
3-Methyl-2-oxazolidone
Formule:
C₄H₇NO₂ Kookpunt: 88°C |
MDL Nummer:
MFCD00009759 CAS nummer: 19836-78-3 EINECS: 243-361-1 |
|
Specification Test Results
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...