Borane-morpholine complex ≥95.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

M0898-25G M0898-5G
TCIAM0898-25GEA 123 EUR
TCIAM0898-25G TCIAM0898-5G
Borane-morpholine complex ≥95.0% (by titrimetric analysis)
Borane-morpholine complex
Formule: C₄H₁₂BNO
MDL Nummer: MFCD00066736
CAS nummer: 4856-95-5
EINECS: 225-450-7
UN: 2921
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Potassium iodate Method) min. 95.0 %
Melting point 96.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR