2-(2-Hydroxyethyl)-1-methylpyrrolidine ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
1-Methyl-2-pyrrolidineethanol
M0908-10ML
M0908-25ML
TCIAM0908-10MLEA
73.7
EUR
TCIAM0908-10ML
TCIAM0908-25ML
2-(2-Hydroxyethyl)-1-methylpyrrolidine ≥98.0% (by GC, titration analysis)
2-(2-Hydroxyethyl)-1-methylpyrrolidine
Formule:
C₇H₁₅NO Kookpunt: 111°C Vlampunt: 84°C |
MDL Nummer:
MFCD00003174 CAS nummer: 67004-64-2 EINECS: 266-538-5 |
Specification Test Results
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...