1-(4-Methylphenoxy)-2-propanol ≥90.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M1065-25G
M1065-5G
TCIAM1065-25GEA
212
EUR
TCIAM1065-25G
TCIAM1065-5G
1-(4-Methylphenoxy)-2-propanol ≥90.0% (by GC)
1-(4-Methylphenoxy)-2-propanol
Formule:
C₁₀H₁₄O₂ Kookpunt: 130°C |
MDL Nummer:
MFCD00191544 CAS nummer: 4369-08-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...