Monocaprin ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Glycerol alpha-Monodecanoate, 1-Monodecanoyl Glycerol
M1072-1G
M1072-5G
TCIAM1072-1GEA
59.4
EUR
TCIAM1072-1G
TCIAM1072-5G
Monocaprin ≥98.0% (by GC)
Monocaprin
Formule:
C₁₃H₂₆O₄ MW: 246.35 g/mol |
MDL Nummer:
MFCD00056656 CAS nummer: 26402-22-2 EINECS: 247-667-6 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 51.0 to 55.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...