4-Methylcyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
1-Amino-4-methylcyclohexane
M1117-25ML
M1117-5ML
TCIAM1117-25MLEA
121
EUR
TCIAM1117-25ML
TCIAM1117-5ML
4-Methylcyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥99.0% (by GC)
4-Methylcyclohexylamine (cis- and trans- mixture)
Formule:
C₇H₁₅N MW: 113.20 g/mol Kookpunt: 152°C Vlampunt: 27°C |
MDL Nummer:
MFCD00001495 CAS nummer: 6321-23-9 EINECS: 228-673-8 UN: 2734 ADR: 8 II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...