4-Methoxyphenylphosphonic dichloride ≥90.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M1156-5G M1156-1G
TCIAM1156-5GEA 142 EUR
TCIAM1156-5G TCIAM1156-1G
4-Methoxyphenylphosphonic dichloride ≥90.0% (by GC)
4-Methoxyphenylphosphonic dichloride
Formule: C₇H₇Cl₂O₂P
MDL Nummer: MFCD00123296
CAS nummer: 37632-18-1
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d0695947d0fdb4aea411b07120af05223173c04&vid=6d0695947d0fdb4aea411b07120af05223173c04&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 90.0 %