3-((4-Nitrobenzyl)oxy)-3-oxopropanoic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Mono-4-nitrobenzyl Malonate, Malonic acid mono-4-nitrobenzyl ester, 4-Nitrobenzyl Hydrogen Malonate

M1177-25G M1177-5G
TCIAM1177-25GEA 179 EUR
TCIAM1177-25G TCIAM1177-5G
3-((4-Nitrobenzyl)oxy)-3-oxopropanoic acid ≥98.0% (by titrimetric analysis)
3-((4-Nitrobenzyl)oxy)-3-oxopropanoic acid
Formule: C₁₀H₉NO₆
MW: 239.1816 g/mol
MDL Nummer: MFCD00191556
CAS nummer: 77359-11-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 108.0 to 111.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR