(S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine ≥97.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
M1183-1G
M1183-5G
TCIAM1183-1GEA
95.7
EUR
TCIAM1183-1G
TCIAM1183-5G
(S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine ≥97.0% (by GC, titration analysis)
(S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine
Formule:
C₁₁H₂₂N₂ Kookpunt: 126°C Vlampunt: 105°C |
MDL Nummer:
MFCD00077821 CAS nummer: 84466-85-3 |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Specific rotation [a]20/D | -67.0 to -71.0 deg (C=0.5, EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...