S-Methyl methanethiosulphonate (MMTS) ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
M1382-25G
M1382-5G
M1382-100G
TCIAM1382-25GEA
273
EUR
TCIAM1382-25G
TCIAM1382-5G
TCIAM1382-100G
S-Methyl methanethiosulphonate (MMTS) ≥97.0% (by GC)
S-Methyl methanethiosulphonate (MMTS)
Formule:
C₂H₆O₂S₂ Kookpunt: 125°C Vlampunt: 87°C |
MDL Nummer:
MFCD00007565 CAS nummer: 2949-92-0 EINECS: 220-970-0 UN: 2810 ADR: 6.1,III |
Specification Test Results
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...