7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene ≥95.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: MTBD, 1,3,4,6,7,8-Hexahydro-1-methyl-2H-pyrimido[1,2-a]pyrimidine

M1443-1G M1443-5G
TCIAM1443-1GEA 52.8 EUR
TCIAM1443-1G TCIAM1443-5G
7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene ≥95.0% (by GC, titration analysis)
7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene
Formule: C₈H₁₅N₃
MW: 153.23 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00043004
CAS nummer: 84030-20-6
EINECS: 281-791-1
UN: 3267
ADR: 8 II

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR