3-(Methoxymethoxy)benzaldehyde ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M1448-25G M1448-5G
TCIAM1448-25GEA 351 EUR
TCIAM1448-25G TCIAM1448-5G
3-(Methoxymethoxy)benzaldehyde ≥95.0% (by GC)
3-(Methoxymethoxy)benzaldehyde
Formule: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Nummer: MFCD04038425
CAS nummer: 13709-05-2
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %