4-Methoxyphenyl-2,3,4,6-tetra-O-acetyl-β-D-galactopyranoside ≥98.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
(2R,3S,4S,5R,6S)-2-(Acetoxymethyl)-6-(4-methoxyphenoxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate
M1477-25G
M1477-5G
TCIAM1477-25GEA
159
EUR
TCIAM1477-25G
TCIAM1477-5G
4-Methoxyphenyl-2,3,4,6-tetra-O-acetyl-β-D-galactopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-2,3,4,6-tetra-O-acetyl-beta-D-galactopyranoside
Formule:
C₂₁H₂₆O₁₁ MW: 454.43 g/mol Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD06797123 CAS nummer: 2872-65-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 101.0 to 105.0 deg-C |
Specific rotation [a]20/D | +2.0 to +4.0 deg (C=1, CHCl3) |
Solubility in Chloroform | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...