2-Methyl-1H-indole-3-carbaldehyde ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-Methylindole-3-carboxaldehyde, 3-Formyl-2-methylindole
M1485-25G M1485-5G
TCIAM1485-25GEA 255 EUR
TCIAM1485-25G TCIAM1485-5G
2-Methyl-1H-indole-3-carbaldehyde ≥98.0% (by GC)
2-Methyl-1H-indole-3-carbaldehyde
Formule: C₁₀H₉NO
MW: 159.1846 g/mol
MDL Nummer: MFCD00012077
CAS nummer: 5416-80-8
EINECS: 226-512-6
Order Now
Specification Test Results
Purity (GC) min. 98.0 %
Melting point 201.0 to 205.0 deg-C