2-Methyl-1,3-propanediamine ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
1,3-Diamino-2-methylpropane
M1657-1G
M1657-5G
TCIAM1657-1GEA
102
EUR
TCIAM1657-1G
TCIAM1657-5G
2-Methyl-1,3-propanediamine ≥98.0% (by GC, titration analysis)
2-Methyl-1,3-propanediamine
Formule:
C₄H₁₂N₂ |
CAS nummer:
2400-78-4 UN: 2734 ADR: 8,II |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...