Methyl-3-(4-hydroxy-3-methoxyphenyl)propionate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M1803-25G M1803-5G
TCIAM1803-25GEA 578 EUR
TCIAM1803-25G TCIAM1803-5G
Methyl-3-(4-hydroxy-3-methoxyphenyl)propionate ≥98.0% (by GC)
Methyl-3-(4-hydroxy-3-methoxyphenyl)propionate
Formule: C₁₁H₁₄O₄
Kookpunt: 146°C
MDL Nummer: MFCD11975434
CAS nummer: 56024-44-3

Order Now

Specification Test Results

Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR