1-Methyl-L-glutamate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M1861-5G M1861-1G
TCIAM1861-5GEA 124 EUR
TCIAM1861-5G TCIAM1861-1G
1-Methyl-L-glutamate ≥98.0% (by titrimetric analysis)
1-Methyl-L-glutamate
Formule: C₆H₁₁NO₄
Smeltpunt: 145°C
MDL Nummer: MFCD00171677
CAS nummer: 6384-08-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca705ff628fe8126e5ccb82476d620024db1de98&vid=7a420d75eb85bcac66135e3705bec1c21e3faa57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +34.0 to +37.0 deg (C=2, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR