2-Methyl-1-phenyl-2-butanol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M2010-25G M2010-5G
TCIAM2010-25GEA 112 EUR
TCIAM2010-25G TCIAM2010-5G
2-Methyl-1-phenyl-2-butanol ≥98.0% (by GC)
2-Methyl-1-phenyl-2-butanol
Formule: C₁₁H₁₆O
Kookpunt: 61°C
MDL Nummer: MFCD01708473
CAS nummer: 772-46-3
EINECS: 212-249-4
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %