3-Methyl-1-phenyl-3-pentanol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M2011-25G M2011-5G
TCIAM2011-25GEA 37.4 EUR
TCIAM2011-25G TCIAM2011-5G
3-Methyl-1-phenyl-3-pentanol ≥98.0% (by GC)
3-Methyl-1-phenyl-3-pentanol
Formule: C₁₂H₁₈O
Kookpunt: 130°C
MDL Nummer: MFCD00021825
CAS nummer: 10415-87-9
EINECS: 233-889-0
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %