Methyl heptafluoroisobutyrate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Heptafluoroisobutyric Acid Methyl Ester

M2022-25G M2022-5G
TCIAM2022-25GEA 373 EUR
TCIAM2022-25G TCIAM2022-5G
Methyl heptafluoroisobutyrate ≥98.0% (by GC)
Methyl heptafluoroisobutyrate
Formule: C₅H₃F₇O₂
Kookpunt: 77°C
MDL Nummer: MFCD00633403
CAS nummer: 680-05-7
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR