O-(2-Methoxyisopropyl)hydroxylamine ≥95.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

M2085-1G M2085-5G
TCIAM2085-1GEA 131 EUR
TCIAM2085-1G TCIAM2085-5G
O-(2-Methoxyisopropyl)hydroxylamine ≥95.0% (by GC, titration analysis)
O-(2-Methoxyisopropyl)hydroxylamine
Formule: C₄H₁₁NO₂
Kookpunt: 73°C
Bewaar temperatuur: Koelkast
CAS nummer: 103491-33-4
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR