6-Methyl-4-phenylchroman-2-one ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M2093-25G
M2093-5G
TCIAM2093-25GEA
152
EUR
TCIAM2093-25G
TCIAM2093-5G
6-Methyl-4-phenylchroman-2-one ≥98.0% (by GC)
6-Methyl-4-phenylchroman-2-one
Formule:
C₁₆H₁₄O₂ Kookpunt: 230°C |
MDL Nummer:
MFCD03695019 CAS nummer: 40546-94-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 79.0 to 84.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...