4-[(4-Methyl-1-piperazinyl)methyl]benzoic acid dihydrochloride ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 4-[(4-Methylpiperazin-1-yl)methyl]benzoic acid dihydrochloride

M2110-25G M2110-5G
TCIAM2110-25GEA 288 EUR
TCIAM2110-25G TCIAM2110-5G
4-[(4-Methyl-1-piperazinyl)methyl]benzoic acid dihydrochloride ≥98.0% (by HPLC)
4-[(4-Methyl-1-piperazinyl)methyl]benzoic acid dihydrochloride
Formule: C₁₃H₁₈N₂O₂
MW: 307.22 g/mol
MDL Nummer: MFCD07772867
CAS nummer: 106261-49-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR