1-Methylimidazole-2-carbaldehyde ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-Methyl-1H-imidazole-2-carbaldehyde, 1-Methylimidazole-2-carboxaldehyde
M2176-1G M2176-5G
TCIAM2176-1GEA 38.5 EUR
TCIAM2176-1G TCIAM2176-5G
1-Methylimidazole-2-carbaldehyde ≥98.0% (by GC)
1-Methylimidazole-2-carbaldehyde
Formule: C₅H₆N₂O
Kookpunt: 74°C
Smeltpunt: 39°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD01321308
CAS nummer: 13750-81-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af0dba0e962791fe464afd847b98506fad486737&vid=af0dba0e962791fe464afd847b98506fad486737&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Acid Value max. 5.0