6-Methyl-4-chromanone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M2249-1G M2249-5G
TCIAM2249-1GEA 38.5 EUR
TCIAM2249-1G TCIAM2249-5G
6-Methyl-4-chromanone ≥98.0% (by GC)
6-Methyl-4-chromanone
Formule: C₁₀H₁₀O₂
Kookpunt: 162°C
MDL Nummer: MFCD00024071
CAS nummer: 39513-75-2

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 33.0 to 37.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR