Methyl-3-hydroxy-4-nitrobenzoate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M2250-5G M2250-25G
TCIAM2250-5GEA 37.4 EUR
TCIAM2250-5G TCIAM2250-25G
Methyl-3-hydroxy-4-nitrobenzoate ≥98.0% (by GC)
Methyl-3-hydroxy-4-nitrobenzoate
Formule: C₈H₇NO₅
MDL Nummer: MFCD00016997
CAS nummer: 713-52-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83797d53cbf755404cfc4b15c9676ccf0712a3c6&vid=83797d53cbf755404cfc4b15c9676ccf0712a3c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point gt. 90.0 to 94.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR