2'-O-Methyluridine ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
1-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 2′-O-Methyluridine
M2290-1G
M2290-5G
TCIAM2290-1GEA
50.6
EUR
TCIAM2290-1G
TCIAM2290-5G
2'-O-Methyluridine ≥98.0% (by HPLC, titration analysis)
2'-O-Methyluridine
Formule:
C₁₀H₁₄N₂O₆ MW: 258.23 g/mol Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00056054 CAS nummer: 2140-76-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 159.0 to 163.0 deg-C |
Specific rotation [a]20/D | +40.0 to +44.0 deg (C=2、H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...