1-Methyl-3-[6-(methylthio)hexyl]imidazolium p-toluenesulphonate ≥95.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M2321-1G M2321-5G
TCIAM2321-1GEA 138 EUR
TCIAM2321-1G TCIAM2321-5G
1-Methyl-3-[6-(methylthio)hexyl]imidazolium p-toluenesulphonate ≥95.0% (by HPLC, total nitrogen)
1-Methyl-3-[6-(methylthio)hexyl]imidazolium p-toluenesulphonate
Formule: C₁₈H₂₈N₂O₃S₂
CAS nummer: 1352947-63-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e65c93de6013ce4fe7994162d08a9331349f7ab2&vid=e65c93de6013ce4fe7994162d08a9331349f7ab2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR