Methyl-3,5-diamino-6-chloropyrazine-2-carboxylate ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Methyl 6-chloro-3,5-diaminopyrazine-2-carboxylate, Methyl 3,5-diamino-6-chloropyrazine-2-carboxylate
M2322-5G
M2322-25G
TCIAM2322-5GEA
121
EUR
TCIAM2322-5G
TCIAM2322-25G
Methyl-3,5-diamino-6-chloropyrazine-2-carboxylate ≥98.0% (by GC, titration analysis)
Methyl-3,5-diamino-6-chloropyrazine-2-carboxylate
Formule:
C₆H₇ClN₄O₂ MW: 202.60 g/mol |
MDL Nummer:
MFCD01928388 CAS nummer: 1458-01-1 EINECS: 215-946-1 |
|
Specification Test Results
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 211.0 to 215.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...