2-Methoxy-5-pyrimidineboronic acid (contains varying amounts of Anhydride)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
M2335-1G
M2335-5G
TCIAM2335-1GEA
34.1
EUR
TCIAM2335-1G
TCIAM2335-5G
2-Methoxy-5-pyrimidineboronic acid (contains varying amounts of Anhydride)
2-Methoxy-5-pyrimidineboronic acid
Formule:
C₅H₇BN₂O₃ Smeltpunt: 161°C |
MDL Nummer:
MFCD03094664 CAS nummer: 628692-15-9 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 114.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...