4-Maleimidobutyric acid ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)butanoic acid

M2337-1G M2337-5G
TCIAM2337-1GEA 56.1 EUR
TCIAM2337-1G TCIAM2337-5G
4-Maleimidobutyric acid ≥98.0% (by GC)
4-Maleimidobutyric acid
Formule: C₈H₉NO₄
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00043139
CAS nummer: 57078-98-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9773e6286c4e5948f26fd75dfde0401697d133e5&vid=9773e6286c4e5948f26fd75dfde0401697d133e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 88.0 to 98.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR