Methyl-1,2,4-triazole-3-carboxylate ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M2368-5G M2368-25G
TCIAM2368-5GEA 17.6 EUR
TCIAM2368-5G TCIAM2368-25G
Methyl-1,2,4-triazole-3-carboxylate ≥98.0% (by HPLC, titration analysis)
Methyl-1,2,4-triazole-3-carboxylate
Formule: C₄H₅N₃O₂
MDL Nummer: MFCD00135989
CAS nummer: 4928-88-5
EINECS: 480-470-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ca0bd91dd612eeeb21061b29de19103333f4393&vid=5ca0bd91dd612eeeb21061b29de19103333f4393&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR