1-Methyl-1H-pyrazol-4-carbaldehyde ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M2372-1G M2372-5G
TCIAM2372-1GEA 20.9 EUR
TCIAM2372-1G TCIAM2372-5G
1-Methyl-1H-pyrazol-4-carbaldehyde ≥98.0% (by GC)
1-Methyl-1H-pyrazol-4-carbaldehyde
Formule: C₅H₆N₂O
Kookpunt: 81°C
MDL Nummer: MFCD00460465
CAS nummer: 25016-11-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7e379a5532c925e64a65c6e5345a5a018f28fa9&vid=c7e379a5532c925e64a65c6e5345a5a018f28fa9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 31.0 to 35.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR