N-Methylisopropylamine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: N-Isopropylmethylamine

M2429-5ML M2429-25ML
TCIAM2429-5MLEA 39.6 EUR
TCIAM2429-5ML TCIAM2429-25ML
N-Methylisopropylamine ≥98.0% (by GC)
N-Methylisopropylamine
Formule: C₄H₁₁N
MW: 73.14 g/mol
Kookpunt: 50°C
Vlampunt: –32°C
MDL Nummer: MFCD00042859
CAS nummer: 4747-21-1
EINECS: 225-266-7
UN: 2734
ADR: 8 I

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR