Moroxydine hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M2443-5G M2443-25G
TCIAM2443-5GEA 49.5 EUR
TCIAM2443-5G TCIAM2443-25G
Moroxydine hydrochloride ≥98.0% (by HPLC, titration analysis)
Moroxydine hydrochloride
Formule: C₆H₁₃N₅O
MDL Nummer: MFCD00012798
CAS nummer: 3160-91-6
EINECS: 221-612-6
Merck Index: 14,06272
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4f1ca8ff4db52e81661e43f77e72f4d6111debb&vid=c4f1ca8ff4db52e81661e43f77e72f4d6111debb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 200.0 to 204.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR