2-(4-Methyl-2-thioxo-2,3-dihydrothiazol-5-yl)acetic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
5-(Carboxymethyl)-2-mercapto-4-methylthiazole, (2-Mercapto-4-methyl-5-thiazolyl)acetic acid
M2481-5G
M2481-25G
TCIAM2481-5GEA
41.8
EUR
TCIAM2481-5G
TCIAM2481-25G
2-(4-Methyl-2-thioxo-2,3-dihydrothiazol-5-yl)acetic acid ≥98.0% (by GC, titration analysis)
2-(4-Methyl-2-thioxo-2,3-dihydrothiazol-5-yl)acetic acid
Formule:
C₆H₇NO₂S₂ MW: 189.2553 g/mol |
MDL Nummer:
MFCD01750430 CAS nummer: 34272-64-5 EINECS: 251-905-4 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...