3-Mercaptohexyl acetate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M2497-5G M2497-25G
TCIAM2497-5GEA 52.8 EUR
TCIAM2497-5G TCIAM2497-25G
3-Mercaptohexyl acetate ≥97.0% (by GC)
3-Mercaptohexyl acetate
Formule: C₈H₁₆O₂S
MDL Nummer: MFCD00792516
CAS nummer: 136954-20-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2a5c802c4a9dcdbc7b1819d343ba3261113fc80&vid=a2a5c802c4a9dcdbc7b1819d343ba3261113fc80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR