4-Methylpiperazine-1-ethanol ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-(2-Hydroxyethyl)-4-methylpiperazine
M2499-25G M2499-5G
TCIAM2499-25GEA 161 EUR
TCIAM2499-25G TCIAM2499-5G
4-Methylpiperazine-1-ethanol ≥98.0% (by GC, titration analysis)
4-Methylpiperazine-1-ethanol
Formule: C₇H₁₆N₂O
MW: 144.22 g/mol
Kookpunt: 55°C
MDL Nummer: MFCD03840691
CAS nummer: 5464-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d4b20db0b3b52fc73115d8c35f8f0830d14e59d&vid=7d4b20db0b3b52fc73115d8c35f8f0830d14e59d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %