Methyl-4-fluoro-3-nitrobenzoate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Methyl 4-fluoro-3-nitrobenzoate, 4-Fluorobenzoic acid methyl ester

M2535-1G M2535-5G
TCIAM2535-1GEA 27.5 EUR
TCIAM2535-1G TCIAM2535-5G
Methyl-4-fluoro-3-nitrobenzoate ≥98.0% (by GC)
Methyl-4-fluoro-3-nitrobenzoate
Formule: C₈H₆FNO₄
MW: 199.14 g/mol
Kookpunt: 119°C
MDL Nummer: MFCD00196161
CAS nummer: 329-59-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=683a58da1091aa6157a9567def1e7a1d05f7fc87&vid=683a58da1091aa6157a9567def1e7a1d05f7fc87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR