3-Nitro-2-picoline ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
M2548-1G
M2548-5G
TCIAM2548-1GEA
40.7
EUR
TCIAM2548-1G
TCIAM2548-5G
3-Nitro-2-picoline ≥98.0% (by GC)
3-Nitro-2-picoline
Formule:
C₆H₆N₂O₂ Kookpunt: 86°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD04114137 CAS nummer: 18699-87-1 |
|
Specification Test Results
Appearance | Light yellow to Brown powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...