6-(Methoxycarbonyl)picolinic acid ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2,6-Pyridinedicarboxylic acid monomethyl ester, Dipicolinic acid monomethyl ester, Monomethyl 2,6-Pyridinedicarboxylate, Monomethyl dipicolinate

M2580-1G M2580-5G
TCIAM2580-1GEA 101 EUR
TCIAM2580-1G TCIAM2580-5G
6-(Methoxycarbonyl)picolinic acid ≥97.0% (by GC, titration analysis)
6-(Methoxycarbonyl)picolinic acid
Formule: C8H7NO4
MW: 181.1455 g/mol
MDL Nummer: MFCD00806109
CAS nummer: 7170-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0ee01605b7c2811efbb7f27f652e21d98a69107&vid=f0ee01605b7c2811efbb7f27f652e21d98a69107&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR