3-Methyl-1H-pyrazole-5-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M2607-1G M2607-5G
TCIAM2607-1GEA 68.2 EUR
TCIAM2607-1G TCIAM2607-5G
3-Methyl-1H-pyrazole-5-carboxylic acid ≥98.0% (by HPLC, titration analysis)
3-Methyl-1H-pyrazole-5-carboxylic acid
Formule: C₅H₆N₂O₂
Smeltpunt: 245°C
MDL Nummer: MFCD00462235
CAS nummer: 696-22-0
Merck Index: 14,06115
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7059f2d80ded0b9bfa5e50a8ded1886454bc957d&vid=1e5ac4602fa61c077f88ac477b82159ace14fb81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR