[2-[(Methylthio)(2H-pyrrol-2-ylidene)methyl]-1H-pyrrole](difluoroborane) ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
M2609-200MG
M2609-1G
TCIAM2609-200MGEA
112
EUR
TCIAM2609-200MG
TCIAM2609-1G
[2-[(Methylthio)(2H-pyrrol-2-ylidene)methyl]-1H-pyrrole](difluoroborane) ≥98.0% (by GC)
[2-[(Methylthio)(2H-pyrrol-2-ylidene)methyl]-1H-pyrrole](difluoroborane)
Formule:
C₁₀H₉BF₂N₂S |
CAS nummer:
892505-41-8 |
|
Specification Test Results
Appearance | Red to Dark red to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Lambda max. | 486.0 to 490.0 nm (EtOH) |
Molar extinction coefficient | min. 33000 (EtOH, 486.0 to 490.0nm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...